Learn More
Trajan Scientific Needle eVol Medical Lab 5/PK
SDP
Catalog No. 5000493800
Encompass_Preferred
5 cm 0 50 mm OD Waters LC tipped needle for 1-2 5 mL syringes and 500 L eVol syringes PK5
Catalog No. 50-004-93800 Supplier Trajan Scientific Supplier No. 039260
5 cm 0 50 mm OD Waters LC tipped needle for 1-2 5 mL syringes and 500 L eVol syringes PK5