Learn More
Trajan Scientific Needle eVol Medical Lab 5/PK
SDP
Supplier:  Trajan Scientific 039038
Encompass_Preferred
18 cm 0 63 mm OD cone tipped needle for 1-2 5 mL syringes (and 500 L eVol syringes) PK2
Catalog No. 50-004-93674