Learn More
Waters Corp Waters Needle Guide Fitting, LC, Countersunk Hole, Reproducible Injection
Catalog No. 50792699 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This needle guide fitting is designed for use in LC systems, specifically with the 2777 Sample Manager. It is installed in port 1 of the valve and features a wide diameter to mate with the syringe needle guide on the injection unit. The fitting also includes a countersunk hole to facilitate the precise insertion of the syringe needle, ensuring reproducible sample injection and minimizing carryover.
  • Installed in port 1 of the valve
  • Features a wide diameter for syringe needle guide mating
  • Includes a countersunk hole for easy syringe needle insertion
  • Ensures reproducible sample injection
  • Minimizes sample carryover
  • Holds the needle seal securely
  • Compatible with LC systems
Catalog No. 50-792-699 Supplier Waters Corp Supplier No. 700002211
This needle guide fitting is designed for use in LC systems, specifically with the 2777 Sample Manager. It is installed in port 1 of the valve and features a wide diameter to mate with the syringe needle guide on the injection unit. The fitting also includes a countersunk hole to facilitate the precise insertion of the syringe needle, ensuring reproducible sample injection and minimizing carryover.
  • Installed in port 1 of the valve
  • Features a wide diameter for syringe needle guide mating
  • Includes a countersunk hole for easy syringe needle insertion
  • Ensures reproducible sample injection
  • Minimizes sample carryover
  • Holds the needle seal securely
  • Compatible with LC systems