Learn More
Agilent Technologies NEEDLE HDSPACE TRANSFER LINE
Catalog No. NC2081545
Encompass
Needle, headspace transfer line, deactivated, 0.7 mm od, 0.4 mm id
Catalog No. NC2081545 Supplier Agilent Technologies Supplier No. 2322590005
Needle, headspace transfer line, deactivated, 0.7 mm od, 0.4 mm id