Learn More
Med Vet International Miltex Olsen-Hegar Needle Holder, 6.5in., Tungsten Carbide
Catalog No. 502832135
Encompass_Preferred
The Miltex MH8-16TC Olsen-Hegar Needle Holder have a length of 6.5in.. These Olsen-Hegar Needle holders are made of German grade stainless steel and Tungsten Carbide. They are a convenient surgical instruments as they have a blade at the end of the jaws.
Catalog No. 50-283-2135 Supplier Med Vet International Supplier No. MH816TC
The Miltex MH8-16TC Olsen-Hegar Needle Holder have a length of 6.5in.. These Olsen-Hegar Needle holders are made of German grade stainless steel and Tungsten Carbide. They are a convenient surgical instruments as they have a blade at the end of the jaws.