Learn More
Fine Science Tools Round Handled Needle Holders, 12.5cm
SDP
Catalog No. NC0410033
Encompass
Product: Round Handled Needle Holder - Straight/Smooth/12.5cm/with Lock/ Tip Shape: Straight/ Tip Width: 0.6mm/ Clamping Length: 7mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12.5cm/ Serrated: No/ Unit: Each
Catalog No. NC0410033 Supplier Fine Science Tools Supplier No. 1207512
Product: Round Handled Needle Holder - Straight/Smooth/12.5cm/with Lock/ Tip Shape: Straight/ Tip Width: 0.6mm/ Clamping Length: 7mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12.5cm/ Serrated: No/ Unit: Each