Learn More
Fine Science Tools Olsen-Hegar Needle Holders with Suture Cutters, Tungsten Carbide, 12 cm
SDP
Catalog No. NC9131906
Encompass
Product: Olsen-Hegar Needle Holder with Scissors/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 7mm/ Scissors: Yes/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12cm/ Serrated: Yes/ Feature: Tungsten Carbide/ Unit: Each
Catalog No. NC9131906 Supplier Fine Science Tools Supplier No. 1250212
Product: Olsen-Hegar Needle Holder with Scissors/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 7mm/ Scissors: Yes/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12cm/ Serrated: Yes/ Feature: Tungsten Carbide/ Unit: Each