Learn More
World Precision Instrument Halsey Needle Holder, 14cm, Straight, Serrated, German
SDP
Catalog No. 50822214
Encompass
Halsey needle holders are used to hold and manipulate surgical needles during procedures. Halsey needle holders have a scissor-like jaw design with a flat, blunt tip. They have a locking mechanism to secure the jaws as they grasp the needle firmly. While Halsey needle holders are available in sizes from 4 to 7 inches, these are 5.5" long. Choose the size of your needle holders (also called needle drivers) based on the size of your needles. 
Catalog No. 50-822-214 Supplier World Precision Instrument Supplier No. 14110G
Halsey needle holders are used to hold and manipulate surgical needles during procedures. Halsey needle holders have a scissor-like jaw design with a flat, blunt tip. They have a locking mechanism to secure the jaws as they grasp the needle firmly. While Halsey needle holders are available in sizes from 4 to 7 inches, these are 5.5" long. Choose the size of your needle holders (also called needle drivers) based on the size of your needles. 