Learn More
Roboz Surgical Needle Holder For Micro Dissecting Needles RS-6063 To RS-6070; Light weight, solid aluminum handle with nickel plated precision chuck; 5 1/4" Long.
SDP
Catalog No. NC2892261
Encompass
Needle Holder For Micro Dissecting Needles RS-6063 To RS-6070; Light weight, solid aluminum handle with nickel plated precision chuck; 5 1/4" Long.
Catalog No. NC2892261 Supplier Roboz Surgical Supplier No. RS6060
Needle Holder For Micro Dissecting Needles RS-6063 To RS-6070; Light weight, solid aluminum handle with nickel plated precision chuck; 5 1/4" Long.