Learn More
Med Vet International Miltex Baumgartner Meisterhand Needle Holder, Serrated Jaws, 5.75in, German, Mh8-40
Catalog No. 502830996
Encompass_Preferred
Locking style needle holder holds suturing needle in place during surgical procedures. MeisterHand quality and precision in craftsman ship guarantees excellent build composure. Serrated jaws. 5.75in total length. Made with top of the line, medical-standard German steel.
Catalog No. 50-283-0996 Supplier Med Vet International Supplier No. MILMH840
Add to Cart
Add to Cart
Locking style needle holder holds suturing needle in place during surgical procedures. MeisterHand quality and precision in craftsman ship guarantees excellent build composure. Serrated jaws. 5.75in total length. Made with top of the line, medical-standard German steel.