Learn More
Med Vet International Miltex Meisterhand Brown Needle Holder, Convex Jaws, Fine Serrations, 5.25in, German, Mh8-22
Catalog No. 502830970
Encompass_Preferred
Highly versatile instrument used to hold suture needles or drivers during surgical procedures. Convex jaws with fine serrations for increased grip. MeisterHand build quality ensures the best craftsmanship that is offered. 5.25in total length. Handcrafted with medical grade German steel.
Catalog No. 50-283-0970 Supplier Med Vet International Supplier No. MILMH822
Highly versatile instrument used to hold suture needles or drivers during surgical procedures. Convex jaws with fine serrations for increased grip. MeisterHand build quality ensures the best craftsmanship that is offered. 5.25in total length. Handcrafted with medical grade German steel.