Learn More
Med Vet International Miltex Castroviejo Needle Holder, Straight, Smooth Jaws w/Lock, 5.5in, German, 18-1828Tc
Catalog No. 502829094
Encompass_Preferred
The edges on the side of the straight jaws are chamfered to prevent accidental cutting of suture material when held in the jaws. Smooth finish. Straight with lock. 5.5in total length. Made with top of the line German steel.
Catalog No. 50-282-9094 Supplier Med Vet International Supplier No. MIL181828TC
The edges on the side of the straight jaws are chamfered to prevent accidental cutting of suture material when held in the jaws. Smooth finish. Straight with lock. 5.5in total length. Made with top of the line German steel.