Learn More
Med Vet International Miltex Castroviejo Needle Holder, Straight, Serrated Jaws w/Lock, Tungsten, 5.5in, German, 18-1828A-Tc
Catalog No. 502829093
Encompass_Preferred
With straight needle holder jaws made of tungsten carbide, the hardest steel used in surgical instruments, this instrument ensures prolonged durability and exceptional quality. Serrations on the jaws allow greater grip and prevent slippage. Straight with lock. The edges around the box lock have distinct non-catch bevel. 5.5in total length. Made with German steel.
Catalog No. 50-282-9093 Supplier Med Vet International Supplier No. MIL181828ATC
With straight needle holder jaws made of tungsten carbide, the hardest steel used in surgical instruments, this instrument ensures prolonged durability and exceptional quality. Serrations on the jaws allow greater grip and prevent slippage. Straight with lock. The edges around the box lock have distinct non-catch bevel. 5.5in total length. Made with German steel.