Learn More
Med Vet International Miltex Vantage Castroviejo Needle Holder, 5.5in, Straight, with Catch
Catalog No. 502522279
Encompass_Preferred
The Miltex Vantage V918-1828 Castroviejo needle holder is made from high quality Japanese stainless steel and measures 5.5" in length. It has a spring handle with a lock and straight, smooth jaws.
Catalog No. 50-252-2279 Supplier Med Vet International Supplier No. MILV9181828
The Miltex Vantage V918-1828 Castroviejo needle holder is made from high quality Japanese stainless steel and measures 5.5" in length. It has a spring handle with a lock and straight, smooth jaws.