Learn More
Med Vet International Miltex Vantage Collier Needle Holder, 5in., Fenestrated Jaws
Catalog No. 502829842
Encompass_Preferred
The Miltex Vantage V98-2 Collier needle holder is made from high quality Japanese stainless steel and measures 5" in length. It has a finger ring handle with a ratchet lock and straight, serrated jaws that are fenestrated.
Catalog No. 50-282-9842 Supplier Med Vet International Supplier No. MILV982
The Miltex Vantage V98-2 Collier needle holder is made from high quality Japanese stainless steel and measures 5" in length. It has a finger ring handle with a ratchet lock and straight, serrated jaws that are fenestrated.