Learn More
Med Vet International Miltex Meisterhand Crile-Wood Needle Holder, Serrated Jaws, 5.87in, German, Mh8-50
Catalog No. 502830355
Encompass_Preferred
Crile-Wood needle holders are used to grasp and guide the needle when suturing. The jaws are serrated to provide additional grip. Similar to Mayo-Heger holders but the tip is finer and gently tapered. 5.87in total length. Made with top of the line German steel.
Catalog No. 50-283-0355 Supplier Med Vet International Supplier No. MILMH850
Crile-Wood needle holders are used to grasp and guide the needle when suturing. The jaws are serrated to provide additional grip. Similar to Mayo-Heger holders but the tip is finer and gently tapered. 5.87in total length. Made with top of the line German steel.