Learn More
Med Vet International Miltex Crile-Wood Needle Holder, Tungsten, Straight, Serrated Jaws, 8in, German, 8-52Tc
Catalog No. 502832031
Encompass_Preferred
Crile-Wood needle holders are used to grasp and guide the needle when suturing. Tungsten carbide inlayed material prevents corrosion and abrasions. Straight serrated jaws for increased grip. Similar to Mayo-Heger holders but the tip is finer and gently tapered. 8in total length. Made with top of the line German steel.
Catalog No. 50-283-2031 Supplier Med Vet International Supplier No. MIL852TC
Crile-Wood needle holders are used to grasp and guide the needle when suturing. Tungsten carbide inlayed material prevents corrosion and abrasions. Straight serrated jaws for increased grip. Similar to Mayo-Heger holders but the tip is finer and gently tapered. 8in total length. Made with top of the line German steel.