Learn More
World Precision Instrument Webster Needle Holder, 12.5cm, Straight, Serrated, Extra Delicate, German
SDP
Supplier:  World Precision Instrument 14109G
Encompass
Webster needle holders are surgical instrument use to hold and manipulate suturing needles. While Webster needle holders are often associated with microsurgery and microvascular procedures, they can be used in a variety of surgical contexts where precise needle handling is required. Webster needle holders have fine tips for precise control when grasping needles. The fine tips make it easier to handle and manipulate small and fine needles used in microsurgery and microvascular procedures. We also offer extra fine tips for smaller needles. Like most needle holders, Webster needle holders have a locking handle for a secure hold on the needle, so that it remains stable during suturing, reducing the risk of slipping or accidental needle sticks. The jaws of some WPI Webster needle holders are lightly textured for a solid grip. Others have smooth jaws to minimize tissue damage. The ratchet handle lets you adjust the tension on the needle, ensuring a secure grip while suturing.
Catalog No. 50-822-212