Learn More
World Precision Instrument Derf Needle Holder, 12cm, Straight, Serrated
SDP
Catalog No. 50822599
Encompass
Derf Needle Holders are surgical instruments designed for holding needles during suturing. The small size (12.5 cm/5") and the short jaws are perfect for smaller needles like 5-0 or 6-0 sutures. The tip has fine serrations and a groove in the center. The groove helps to securely hold thicker surgical needle. The tips of needle holders are generally shorter than those of surgical forceps. The interlocking ratchet handle ensures a firm grasp of the needle when the lock is engaged. The jaws are straight. These needle holders are manufactured from surgical grade stainless steel or premium German steel, both of which are autoclavable or suitable for chemical sterilization. Always clean your surgical instrument thoroughly every time you use them.
Catalog No. 50-822-599 Supplier World Precision Instrument Supplier No. 501246
Derf Needle Holders are surgical instruments designed for holding needles during suturing. The small size (12.5 cm/5") and the short jaws are perfect for smaller needles like 5-0 or 6-0 sutures. The tip has fine serrations and a groove in the center. The groove helps to securely hold thicker surgical needle. The tips of needle holders are generally shorter than those of surgical forceps. The interlocking ratchet handle ensures a firm grasp of the needle when the lock is engaged. The jaws are straight. These needle holders are manufactured from surgical grade stainless steel or premium German steel, both of which are autoclavable or suitable for chemical sterilization. Always clean your surgical instrument thoroughly every time you use them.