Learn More
Med Vet International Miltex Derf Needle Holder, 4.75in., Serrated, Carb-N-Sert Tip, 4000 Teeth Per Square inch
Catalog No. 502830566
Encompass_Preferred
The Miltex 8-4TC Derf needle holder is made of OR grade German stainless steel and measures 4.75" in length. It has a gold-plated, finger ring handle with a ratchet lock and serrated, tungsten carbide (Carb-N-Sert) jaws with 4000 teeth per square "
Catalog No. 50-283-0566 Supplier Med Vet International Supplier No. MIL84TC
The Miltex 8-4TC Derf needle holder is made of OR grade German stainless steel and measures 4.75" in length. It has a gold-plated, finger ring handle with a ratchet lock and serrated, tungsten carbide (Carb-N-Sert) jaws with 4000 teeth per square "