Learn More
Med Vet International Miltex Vantage Derf Needle Holder, 4.75in.
Catalog No. 502829425
Encompass_Preferred
The Miltex Vantage V98-4 Derf needle holder is made from high quality Japanese stainless steel and measures 4.75" in length. It has a finger ring handle with a ratchet lock and straight, serrated jaws.
Catalog No. 50-282-9425 Supplier Med Vet International Supplier No. MILV984
The Miltex Vantage V98-4 Derf needle holder is made from high quality Japanese stainless steel and measures 4.75" in length. It has a finger ring handle with a ratchet lock and straight, serrated jaws.