Learn More
Med Vet International Miltex Meisterhand Derf Needle Holder, Tungsten, Serrated, 4000 P/Sq.in., German, Mh8-4Tc
Catalog No. 502830354
Encompass_Preferred
Frequently used in ophthalmic, dental, and plastic surgery procedures. Serrated blades for increased grip. Blunt tips. Straight blades. MeisterHand build guarantees superior quality. 4.75in total length. 4000 teeth per square " Tungsten carbide inlays ensure increased durability and precision. Handcrafted with medical grade German steel.
Catalog No. 50-283-0354 Supplier Med Vet International Supplier No. MILMH84TC
Frequently used in ophthalmic, dental, and plastic surgery procedures. Serrated blades for increased grip. Blunt tips. Straight blades. MeisterHand build guarantees superior quality. 4.75in total length. 4000 teeth per square " Tungsten carbide inlays ensure increased durability and precision. Handcrafted with medical grade German steel.