Learn More
Med Vet International Miltex Halsey Needle Holder, Smooth Jaws, 5in, German, 8-8
Catalog No. 502831472
Encompass_Preferred
Halsey needle holders are most commonly used for holding suture needles during surgical procedures. It features a cross-hatched and reliable non-slip surface that allows firm gripping of needles during surgical procedures. Ideally used for plastic surgery. Put together with medical grade German steel. Smooth jaws for improved grip. 5in total length. Tungsten carbide inlays provide increased durability against corrosion and abrasions.
Catalog No. 50-283-1472 Supplier Med Vet International Supplier No. MIL88
Halsey needle holders are most commonly used for holding suture needles during surgical procedures. It features a cross-hatched and reliable non-slip surface that allows firm gripping of needles during surgical procedures. Ideally used for plastic surgery. Put together with medical grade German steel. Smooth jaws for improved grip. 5in total length. Tungsten carbide inlays provide increased durability against corrosion and abrasions.