Learn More
Med Vet International Miltex Halsey Meisterhand Needle Holder, Smooth Jaws, 5in, German, Mh8-8
Catalog No. 502830356
Encompass_Preferred
Halsey needle holders are most commonly used for holding suture needles during surgical procedures. It features a cross-hatched and reliable non-slip surface that allows firm gripping of needles during surgical procedures. Ideally used for plastic surgery. Made to match MeisterHand quality specifications. Put together with medical grade German steel. Smooth jaws. 5in total length.
Catalog No. 50-283-0356 Supplier Med Vet International Supplier No. MILMH88
Halsey needle holders are most commonly used for holding suture needles during surgical procedures. It features a cross-hatched and reliable non-slip surface that allows firm gripping of needles during surgical procedures. Ideally used for plastic surgery. Made to match MeisterHand quality specifications. Put together with medical grade German steel. Smooth jaws. 5in total length.