Learn More
Med Vet International Miltex Heaney Needle Holder, Curved w/Lock, Tungsten, Serrated Jaws, 8in, 2600 Teeth P/Sq. in., German, 8-100Tc
Catalog No. 502830671
Encompass_Preferred
Ratcheted finger-ring instrument utilized to hold light-to-medium weight needles while suturing. Tungsten carbide inlays ensure the most in durability and precision. The curved jaws and cross-serrated finish are ideal for optimizing grip. Locks come on the jaws for security. 8in total length. Made with German steel.
Catalog No. 50-283-0671 Supplier Med Vet International Supplier No. MIL8100TC
Ratcheted finger-ring instrument utilized to hold light-to-medium weight needles while suturing. Tungsten carbide inlays ensure the most in durability and precision. The curved jaws and cross-serrated finish are ideal for optimizing grip. Locks come on the jaws for security. 8in total length. Made with German steel.