Learn More
Med Vet International Miltex Vantage Mathieu Needle Holder, 5.5in., Spring Handles
Catalog No. 502829704
Encompass_Preferred
The Miltex Vantage V98-28 Mathieu needle holder is made from high quality Japanese stainless steel and measures 5.5" in length. It has spring handles and straight, serrated jaws.
Catalog No. 50-282-9704 Supplier Med Vet International Supplier No. MILV9828
The Miltex Vantage V98-28 Mathieu needle holder is made from high quality Japanese stainless steel and measures 5.5" in length. It has spring handles and straight, serrated jaws.