Learn More
Med Vet International Miltex Mathieu Needle Holder, Serrated Jaws, Tungsten, 2600 Teeth P/Sq.in., 6.25in, German, 8-30Tc
Catalog No. 502830562
Encompass_Preferred
Needle holders that have a pliers handle, a spring leaf and a ratchet at the bottom. The holder can be locked and unlocked by squeezing the handles. Used for suturing and surgical procedures. Tungsten carbide inlays provide increased durability and precision. Jaws are complete with a serrated finished for increased grip. 2600 teeth per square " 6.25in total length. Made with medical grade German steel.
Catalog No. 50-283-0562 Supplier Med Vet International Supplier No. MIL830TC
Needle holders that have a pliers handle, a spring leaf and a ratchet at the bottom. The holder can be locked and unlocked by squeezing the handles. Used for suturing and surgical procedures. Tungsten carbide inlays provide increased durability and precision. Jaws are complete with a serrated finished for increased grip. 2600 teeth per square " 6.25in total length. Made with medical grade German steel.