Learn More
World Precision Instrument Mathieu Fine Needle Holder, TC insert, 14cm, Serrated
SDP
Catalog No. 50822397
Encompass
Mathieu Fine Needle Holders are a precision surgical instrument for microsurgery. They were designed for use in dentistry and oral surgery by French surgeon Jules-Émile Péan Mathieu. Their unique design features a ratchet lock mechanism in the handle, and fine, slender, serrated tips. You will also notice the ring handles are replaced with ergonomic, spring-style handles. The ratchet lock allows the operator to have a light touch with a secure hold for manipulating a suture needle without having to apply constant pressure on the handles. The design of the locking mechanism reduces hand fatigue. Conventional needle holders have ring handles or spring handles. Instead of finger rings or traditional spring handles, Mathieu Needle Holders have curved and elongated handles that are designed to be held between the thumb and ring finger. This design provides a more ergonomic grip and better control over the instrument.
Catalog No. 50-822-397 Supplier World Precision Instrument Supplier No. 500225
Mathieu Fine Needle Holders are a precision surgical instrument for microsurgery. They were designed for use in dentistry and oral surgery by French surgeon Jules-Émile Péan Mathieu. Their unique design features a ratchet lock mechanism in the handle, and fine, slender, serrated tips. You will also notice the ring handles are replaced with ergonomic, spring-style handles. The ratchet lock allows the operator to have a light touch with a secure hold for manipulating a suture needle without having to apply constant pressure on the handles. The design of the locking mechanism reduces hand fatigue. Conventional needle holders have ring handles or spring handles. Instead of finger rings or traditional spring handles, Mathieu Needle Holders have curved and elongated handles that are designed to be held between the thumb and ring finger. This design provides a more ergonomic grip and better control over the instrument.