Learn More
Med Vet International Miltex Mayo-Hegar Needle Holder, 6in.
Catalog No. 502830564
Encompass_Preferred
The Miltex 8-44 Mayo-Hegar Needle Holder are used in a variety of surgical procedures. Mayo-Hegar needle holders are one of the most commonly used surgical instruments for both human and veterinary procedures. These needle holders are made of German grade stainless steel and a length of 6in..
Catalog No. 50-283-0564 Supplier Med Vet International Supplier No. MIL844
The Miltex 8-44 Mayo-Hegar Needle Holder are used in a variety of surgical procedures. Mayo-Hegar needle holders are one of the most commonly used surgical instruments for both human and veterinary procedures. These needle holders are made of German grade stainless steel and a length of 6in..