Learn More
Med Vet International Miltex Mayo-Hegar Quick Release Needle Holder, 7in., Tungsten Carbide
Catalog No. 502832290
Encompass_Preferred
The Miltex 8-746TC Mayo-Hegar Needle Holder are used in a variety of surgical procedures. Mayo-Hegar needle holders are one of the most commonly used surgical instruments for both human and veterinary procedures. These needle holders are made of German grade stainless steel and tungsten carbide and a length of 7in..
Catalog No. 50-283-2290 Supplier Med Vet International Supplier No. MIL8746TC
The Miltex 8-746TC Mayo-Hegar Needle Holder are used in a variety of surgical procedures. Mayo-Hegar needle holders are one of the most commonly used surgical instruments for both human and veterinary procedures. These needle holders are made of German grade stainless steel and tungsten carbide and a length of 7in..