Learn More
Med Vet International Miltex Mayo-Hegar Needle Holder, 5-1/8in., Tungsten Carbide
Catalog No. 502830563
Encompass_Preferred
The Miltex 8-42TC Mayo-Hegar Needle Holders are used in a variety of surgical procedures. Mayo-Hegar needle holders are one of the most commonly used surgical instruments for both human and veterinary procedures. These needle holders are made of German grade stainless steel and tungsten carbide and a length of 5in.
Catalog No. 50-283-0563 Supplier Med Vet International Supplier No. MIL842TC
The Miltex 8-42TC Mayo-Hegar Needle Holders are used in a variety of surgical procedures. Mayo-Hegar needle holders are one of the most commonly used surgical instruments for both human and veterinary procedures. These needle holders are made of German grade stainless steel and tungsten carbide and a length of 5in.