Learn More
Med Vet International Miltex Mayo-Hegar Needle Holder, Serrated, 10.5in, Tungsten, 2600 Teeth, German, 8-130Tc
Catalog No. 502830567
Encompass_Preferred
Versatile in it’s design, Mayo-Hegar needle holders are frequently used in both human and animal procedures. Serrated jaws to allow greater grip on items held. Tungsten carbide coating helps prevent corrosion and abrasions. 10.5in total length. 2600 teeth per square in. Made with top quality German steel.
Catalog No. 50-283-0567 Supplier Med Vet International Supplier No. MIL8130TC
Versatile in it’s design, Mayo-Hegar needle holders are frequently used in both human and animal procedures. Serrated jaws to allow greater grip on items held. Tungsten carbide coating helps prevent corrosion and abrasions. 10.5in total length. 2600 teeth per square in. Made with top quality German steel.