Learn More
Med Vet International Miltex Microsurgery Needle Holder, Round Handles, Curved w/Locks, 5.25in, German, 17-1020
Catalog No. 502830368
Encompass_Preferred
Uniquely designed to allow suturing wounds and incisions with a very precise and smooth execution, reducing the risk of damaging peripheral tissue. Molded with round handles and a curved jaw, complete with a lock. 5.25in total length. Made with medical grade German steel.
Catalog No. 50-283-0368 Supplier Med Vet International Supplier No. MIL171020
Uniquely designed to allow suturing wounds and incisions with a very precise and smooth execution, reducing the risk of damaging peripheral tissue. Molded with round handles and a curved jaw, complete with a lock. 5.25in total length. Made with medical grade German steel.