Learn More
Med Vet International Miltex Meisterhand Olsen-Hegar Needle Holder, Serrated, Suture, 4.75in, German, Mh8-14
Catalog No. 502830966
Encompass_Preferred
Needle holder with scissors that allow you to cut and suture with the same instrument. Intended for 0 suture and finer. Serrated jaw for increased grip. MeisterHand build quality for the best quality offered. 4.75in total length. Handcrafted with medical grade German steel.
Catalog No. 50-283-0966 Supplier Med Vet International Supplier No. MILMH814
Needle holder with scissors that allow you to cut and suture with the same instrument. Intended for 0 suture and finer. Serrated jaw for increased grip. MeisterHand build quality for the best quality offered. 4.75in total length. Handcrafted with medical grade German steel.