Learn More
Med Vet International Miltex Olsen-Hegar Needle Holder, 5-1/2in., Tungsten Carbide
Catalog No. 502830570
Encompass_Preferred
The Miltex 8-15TC Olsen-Hegar Needle Holder have a length of 5.5in.. These Olsen-Hegar Needle holders are made of German grade stainless steel and Tungsten Carbide. They are a convenient surgical instruments as they have a blade at the end of the jaws.
Catalog No. 50-283-0570 Supplier Med Vet International Supplier No. MIL815TC
The Miltex 8-15TC Olsen-Hegar Needle Holder have a length of 5.5in.. These Olsen-Hegar Needle holders are made of German grade stainless steel and Tungsten Carbide. They are a convenient surgical instruments as they have a blade at the end of the jaws.