Learn More
Fine Science Tools Olsen-Hegar Needle Holders with Suture Cutters, 12 cm
SDP
Catalog No. NC9700894
Encompass
Product: Olsen-Hegar Needle Holder with Scissors - Straight/Serrated/12cm/with Lock/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 8.5mm/ Scissors: Yes/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12cm/ Serrated: Yes/ Unit: Each
Catalog No. NC9700894 Supplier Fine Science Tools Supplier No. 1200212
Product: Olsen-Hegar Needle Holder with Scissors - Straight/Serrated/12cm/with Lock/ Tip Shape: Straight/ Tip Width: 1.5mm/ Clamping Length: 8.5mm/ Scissors: Yes/ Lock: Yes/ Alloy: Stainless Steel/ Length: 12cm/ Serrated: Yes/ Unit: Each