Learn More
World Precision Instrument Olsen-Hegar Needle Holder with Suture Scissors, 14cm
SDP
Catalog No. 50822351
Encompass
Olsen-Hegar Needle Holders with Suture Scissors are a dual purpose surgical instrument, combining the functions of a needle holder and surgical scissors into one instrument. This is ideal for suturing, because you can grasp the needle for stitching and use the integrated scissors for trimming the ends without reaching for a different surgical instrument. Streamline your suturing process and save time. Simultaneously, you can reduce the risk of dropping needles. Designed but the American surgeon Dr. George Olsen and gynecologist Dr. Douglas Hegar, the Olsen-Hegar needle holders are often used in orthopedics, plastic surgery, and any application requiring surturing. Life science researchers doing in vivo animal studies like implanting electrodes would also find this instrument useful. Researchers working with microinjection or microfluidics would also benefit from the dual use of this needle holder/scissors combination.
Catalog No. 50-822-351 Supplier World Precision Instrument Supplier No. 500023
Olsen-Hegar Needle Holders with Suture Scissors are a dual purpose surgical instrument, combining the functions of a needle holder and surgical scissors into one instrument. This is ideal for suturing, because you can grasp the needle for stitching and use the integrated scissors for trimming the ends without reaching for a different surgical instrument. Streamline your suturing process and save time. Simultaneously, you can reduce the risk of dropping needles. Designed but the American surgeon Dr. George Olsen and gynecologist Dr. Douglas Hegar, the Olsen-Hegar needle holders are often used in orthopedics, plastic surgery, and any application requiring surturing. Life science researchers doing in vivo animal studies like implanting electrodes would also find this instrument useful. Researchers working with microinjection or microfluidics would also benefit from the dual use of this needle holder/scissors combination.