Learn More
Fine Science Tools Olsen-Hegar Needle Holders with Suture Cutters, 14 cm
SDP
Catalog No. NC9735637
Encompass
Product: Olsen-Hegar Needle Holder with Scissors - Straight/Serrated/14cm/with Lock/ Tip Shape: Straight/ Tip Width: 2mm/ Clamping Length: 9.5mm/ Scissors: Yes/ Lock: Yes/ Alloy: Stainless Steel/ Length: 14cm/ Serrated: Yes/ Unit: Each
Catalog No. NC9735637 Supplier Fine Science Tools Supplier No. 1200214
Product: Olsen-Hegar Needle Holder with Scissors - Straight/Serrated/14cm/with Lock/ Tip Shape: Straight/ Tip Width: 2mm/ Clamping Length: 9.5mm/ Scissors: Yes/ Lock: Yes/ Alloy: Stainless Steel/ Length: 14cm/ Serrated: Yes/ Unit: Each