Learn More
Med Vet International Miltex Olsen-Hegar Needle Holder, 4-3/4in.
Catalog No. 502830568
Encompass_Preferred
The Miltex 8-14 Olsen-Hegar Needle Holder have a length of 4.75in.. These Olsen-Hegar Needle holders are made of German grade stainless steel and are a convenient surgical instruments as they have a blade at the end of the jaws.
Catalog No. 50-283-0568 Supplier Med Vet International Supplier No. MIL814
The Miltex 8-14 Olsen-Hegar Needle Holder have a length of 4.75in.. These Olsen-Hegar Needle holders are made of German grade stainless steel and are a convenient surgical instruments as they have a blade at the end of the jaws.