Learn More
Med Vet International Miltex Vantage Olsen-Hegar Needle Holder, 5in.
Catalog No. 502829840
Encompass_Preferred
The Miltex Vantage V98-15 Olsen-Hegar Needle Holder have a length of 5in.. These Olsen-Hegar Needle holders are made of mid-grade stainless steel and are a convenient surgical instruments as they have a blade at the end of the jaws.
Catalog No. 50-282-9840 Supplier Med Vet International Supplier No. MILV9815
The Miltex Vantage V98-15 Olsen-Hegar Needle Holder have a length of 5in.. These Olsen-Hegar Needle holders are made of mid-grade stainless steel and are a convenient surgical instruments as they have a blade at the end of the jaws.