Learn More
Med Vet International Miltex Vantage Olsen-Hegar Needle Holder, 6in.
Catalog No. 502829424
Encompass_Preferred
The Miltex Vantage V98-16 Olsen-Hegar Needle Holder have a length of 6in.. These Olsen-Hegar Needle holders are made of mid-grade stainless steel and are a convenient surgical instruments as they have a blade at the end of the jaws.
Catalog No. 50-282-9424 Supplier Med Vet International Supplier No. MILV9816
The Miltex Vantage V98-16 Olsen-Hegar Needle Holder have a length of 6in.. These Olsen-Hegar Needle holders are made of mid-grade stainless steel and are a convenient surgical instruments as they have a blade at the end of the jaws.