Learn More
Med Vet International Miltex Vantage Olsen-Hegar Needle Holder 7-1/2in
Catalog No. 502522259
Encompass_Preferred
The Miltex Vantage V98-17 Olsen-Hegar Needle Holder have a length of 7in.. These Olsen-Hegar Needle holders are made of mid-grade stainless steel and are a convenient surgical instruments as they have a blade at the end of the jaws.
Catalog No. 50-252-2259 Supplier Med Vet International Supplier No. MILV9817
The Miltex Vantage V98-17 Olsen-Hegar Needle Holder have a length of 7in.. These Olsen-Hegar Needle holders are made of mid-grade stainless steel and are a convenient surgical instruments as they have a blade at the end of the jaws.