Learn More
Med Vet International Miltex Olsen-Hegar Needle Holder, Smooth Jaws, Extra Delicate, 4.75in, German, 8-14A
Catalog No. 502830569
Encompass_Preferred
Needle holder with scissors that allow you to cut and suture with the same instrument. Intended for 0 suture and finer. Smooth finish on jaw. Extra delicate finish. Tungsten carbide inlays provide increased durability and precision. 4.75in total length. Handcrafted with medical grade German steel.
Catalog No. 50-283-0569 Supplier Med Vet International Supplier No. MIL814A
Needle holder with scissors that allow you to cut and suture with the same instrument. Intended for 0 suture and finer. Smooth finish on jaw. Extra delicate finish. Tungsten carbide inlays provide increased durability and precision. 4.75in total length. Handcrafted with medical grade German steel.