Learn More
Med Vet International Miltex Ryder Needle Holder, Serrated Jaws, Tungsten, 6in, German, Mh8-103Tc
Catalog No. 502830965
Encompass_Preferred
Ryder needle holders are a ratchet finger ring that features narrow jaws and is commonly used with very small needles in cardiovascular, plastic, and neurosurgical procedures. Serrated laterally. Tungsten carbide cross-serrated tips firmly grip the needle still and assists with keeping it from twisting or moving in any way. 6in total length. Made with German steel.
Catalog No. 50-283-0965 Supplier Med Vet International Supplier No. MILMH8103TC
Ryder needle holders are a ratchet finger ring that features narrow jaws and is commonly used with very small needles in cardiovascular, plastic, and neurosurgical procedures. Serrated laterally. Tungsten carbide cross-serrated tips firmly grip the needle still and assists with keeping it from twisting or moving in any way. 6in total length. Made with German steel.