Learn More
Med Vet International Miltex Ryder Needle Holder, Serrated Jaws, Tungsten, 3600 Teeth P/Sq.in., 5.25in, German, Mh8-101Tc
Catalog No. 502832192
Encompass_Preferred
Ryder needle holders are a ratchet finger ring that features narrow jaws and is commonly used with very small needles in cardiovascular, plastic, and neurosurgical procedures. Serrated laterally. Tungsten carbide cross-serrated tips firmly grip the needle still and assists with keeping it from twisting or moving in any way. 3600 teeth per square " 5.25in total length. Made with German steel.
Catalog No. 50-283-2192 Supplier Med Vet International Supplier No. MILMH8101TC
Ryder needle holders are a ratchet finger ring that features narrow jaws and is commonly used with very small needles in cardiovascular, plastic, and neurosurgical procedures. Serrated laterally. Tungsten carbide cross-serrated tips firmly grip the needle still and assists with keeping it from twisting or moving in any way. 3600 teeth per square " 5.25in total length. Made with German steel.