Learn More
Med Vet International Miltex Ryder-Type Needle Holder, Serrated Jaws, Tungsten, 4000 Teeth P/Sq.in., 9in, German, 8-69Tc
Catalog No. 502832033
Encompass_Preferred
Ryder needle holders are a ratchet finger ring that features narrow jaws and is commonly used with very small needles in cardiovascular, plastic, and neurosurgical procedures. Serrated to prevent slippage. Tungsten carbide cross-serrated tips firmly grip the needle still and assists with keeping it from twisting or moving in any way. Extra delicate. 4000 teeth per square " 9in total length. Made with German steel.
Catalog No. 50-283-2033 Supplier Med Vet International Supplier No. MIL869TC
Ryder needle holders are a ratchet finger ring that features narrow jaws and is commonly used with very small needles in cardiovascular, plastic, and neurosurgical procedures. Serrated to prevent slippage. Tungsten carbide cross-serrated tips firmly grip the needle still and assists with keeping it from twisting or moving in any way. Extra delicate. 4000 teeth per square " 9in total length. Made with German steel.