Learn More
Med Vet International Miltex Webster Needle Holder, 4.75in., Tungsten Carbide
Catalog No. 502832439
Encompass_Preferred
The Miltex 8-71-TC Webster Needle Holder has a length of 4.75in. and is made of German grade stainless steel and Tungsten Carbide. This Webster Needle holder has a gold handle and serrated tips. It is commonly used with small needles and fine suture material during plastic surgery or subcutaneous procedures.
Catalog No. 50-283-2439 Supplier Med Vet International Supplier No. MIL87ATC
The Miltex 8-71-TC Webster Needle Holder has a length of 4.75in. and is made of German grade stainless steel and Tungsten Carbide. This Webster Needle holder has a gold handle and serrated tips. It is commonly used with small needles and fine suture material during plastic surgery or subcutaneous procedures.