Learn More
Med Vet International Miltex Webster Needle Holder, 5in.
Catalog No. 502832032
Encompass_Preferred
The Miltex 8-6 Webster Needle Holder has a length of 5in. and is made of German grade stainless steel. This Webster Needle holder has serrated tips and is commonly used with small needles and fine suture material during plastic surgery or subcutaneous procedures.
Catalog No. 50-283-2032 Supplier Med Vet International Supplier No. MIL86
The Miltex 8-6 Webster Needle Holder has a length of 5in. and is made of German grade stainless steel. This Webster Needle holder has serrated tips and is commonly used with small needles and fine suture material during plastic surgery or subcutaneous procedures.