Learn More
World Precision Instrument Needle Holder with Lock, 20cm, Curved, Smooth Jaws, 0.4mm Tips
SDP
Catalog No. 502539495
Encompass
WPI Needle Holder with lock is a surgical instrument used for microsurgery requiring precise control and manipulation of small needles. These have a 0.4mm rounded tip. Chose either straight or curved tips. The curved needle holders are a little longer. Our Needle Holders feature fine tips for grasping and manipulation of small, delicate needles. These needle holders have a spring action for situations when you need constant pressure on the needle and precision control. These are useful when working under a microscope or in confined spaces. They are an excellent choice for left-handed operators, too, as they are designed for ambidextrous use. Be sure to clean, disinfect, and sterilize your surgical instruments promptly after each use to ensure a long life for your surgical tools. These Needle Holders are available in surgical grade stainless steel, and they are autoclavable.
Catalog No. 50-253-9495 Supplier World Precision Instrument Supplier No. 503354
WPI Needle Holder with lock is a surgical instrument used for microsurgery requiring precise control and manipulation of small needles. These have a 0.4mm rounded tip. Chose either straight or curved tips. The curved needle holders are a little longer. Our Needle Holders feature fine tips for grasping and manipulation of small, delicate needles. These needle holders have a spring action for situations when you need constant pressure on the needle and precision control. These are useful when working under a microscope or in confined spaces. They are an excellent choice for left-handed operators, too, as they are designed for ambidextrous use. Be sure to clean, disinfect, and sterilize your surgical instruments promptly after each use to ensure a long life for your surgical tools. These Needle Holders are available in surgical grade stainless steel, and they are autoclavable.