Learn More
Fine Science Tools NEEDLE HOLDER WITH TUNGSTEN CA
SDP
Catalog No. NC9987509
Encompass
Product: Crile-Wood Needle Holder/ Tip Shape: Straight/ Tip Width: 1.7mm/ Clamping Length: 14.5mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 15cm/ Serrated: Yes/ Feature: Tungsten Carbide/ Unit: Each
Catalog No. NC9987509 Supplier Fine Science Tools Supplier No. 1250315
Product: Crile-Wood Needle Holder/ Tip Shape: Straight/ Tip Width: 1.7mm/ Clamping Length: 14.5mm/ Scissors: No/ Lock: Yes/ Alloy: Stainless Steel/ Length: 15cm/ Serrated: Yes/ Feature: Tungsten Carbide/ Unit: Each